Orders LinkContactUsLinkComponent
Inserts
A 9/1 insert, perforated plate for use in E 935-3/1
7×7 mm perforations, 3 mm ridge, PG 8527, 543×773×1 mm
Katalog # 141-0493
Inserts
Katalog # 141-0493
Leverandør:  
Inserts
Inserts
A 9/1 insert, perforated plate for use in E 935-3/1
7×7 mm perforations, 3 mm ridge, PG 8527, 543×773×1 mm
Discontinued
Product Details & Documents
Specifikationer
  • Beskrivelse:
    A 9/1 insert, perforated plate for use in E 935-3/1
    7×7 mm perforations, 3 mm ridge
  • Seq:
    2
  • B×D×H:
    543×773×1 mm
  • Til:
    PG 8527