Orders LinkContactUsLinkComponent
Inserts
Inserts
E 118 suitable for 38 half petri dishes with Ø 100 mm, equipped with 38 holders, height 70 mm, distance approx. 26 mm
Catalog # 141-0379
Supplier:   MIELE
Inserts
Catalog # 141-0379
Supplier:  MIELE
Inserts
Inserts
E 118 suitable for 38 half petri dishes with Ø 100 mm, equipped with 38 holders, height 70 mm, distance approx. 26 mm
Price
3.870,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    E 118 suitable for 38 half petri dishes with Ø 100 mm, equipped with 38 holders, height 70 mm, distance approx. 26 mm
  • Seq:
    47