Accessories
Magnetic strip ML/2 for automatic mobile unit recognition, PG 8535, PG 8536
Catalog # 141-0435
Supplier: MIELE
Jump to:
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifications
- Beskrivelse:Magnetic strip ML/2 for automatic mobile unit recognition
- Seq:22
- Til:PG 8535, PG 8536