Orders LinkContactUsLinkComponent
Cover spectacles, Squale
Cover spectacles, Squale
Welding, anti-scratch, grå, Shade 5 PC
Katalog # BOLESQUWPCC5
Leverandør:   Bollé Safety
Cover spectacles, Squale
Katalog # BOLESQUWPCC5
Leverandør:  Bollé Safety
Cover spectacles, Squale
Cover spectacles, Squale
Welding, anti-scratch, grå, Shade 5 PC
Price
176,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cover glasses (OTG) give excellent protection for wearers of prescription eyewear. Supplied with adjustable cord.
  • Længdejusterbare nylonstænger
  • Formstøbt linse, pandebeskyttelse og sideskjolda
  • Indirekte ventilation
Vægt: 43 g
 : EN 166, EN 170
Specifikationer
  • Model:
    Welding, anti-scratch
  • Farve:
    grå
  • Linser:
    Shade 5 PC