Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Blower foot switch (foot pedal and power cord), provides a hands-free means to temporarily shut off power to the blower when obtaining sensitive balance readings or manipulating fine powders
Katalog # LBCP5441300
Leverandør:   LABCONCO
Accessories for glove boxes, Protector®
Katalog # LBCP5441300
Leverandør:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Blower foot switch (foot pedal and power cord), provides a hands-free means to temporarily shut off power to the blower when obtaining sensitive balance readings or manipulating fine powders
Price
5.550,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Blower foot switch (foot pedal and power cord), provides a hands-free means to temporarily shut off power to the blower when obtaining sensitive balance readings or manipulating fine powders
  • Seq:
    14