Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Kit with HEPA filter and hose barb canister Ø 13 mm, for placing between the glove box and vacuum pump to protect the pump from particulate contaminants
Katalog # LBCP5441400
Leverandør:   LABCONCO
Accessories for glove boxes, Protector®
Katalog # LBCP5441400
Leverandør:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Kit with HEPA filter and hose barb canister Ø 13 mm, for placing between the glove box and vacuum pump to protect the pump from particulate contaminants
Price
6.730,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Kit with HEPA filter and hose barb canister Ø 13 mm, for placing between the glove box and vacuum pump to protect the pump from particulate contaminants
  • Seq:
    20