Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Replacement HEPA filter, 99,99% efficient on 0,03 micron particulates; for use with 5441400 or 5441403 kit
Katalog # LBCP5441401
Leverandør:   LABCONCO
Accessories for glove boxes, Protector®
Katalog # LBCP5441401
Leverandør:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Replacement HEPA filter, 99,99% efficient on 0,03 micron particulates; for use with 5441400 or 5441403 kit
Price
2.850,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Replacement HEPA filter, 99,99% efficient on 0,03 micron particulates; for use with 5441400 or 5441403 kit
  • Seq:
    22