Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Thimble exhaust connector, Ø 152 mm, epoxy coated steel, for connection to house exhaust or remote blower
Katalog # LBCP5440900
Leverandør:   LABCONCO
Accessories for glove boxes, Protector®
Katalog # LBCP5440900
Leverandør:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Thimble exhaust connector, Ø 152 mm, epoxy coated steel, for connection to house exhaust or remote blower
Price
5.950,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Thimble exhaust connector, Ø 152 mm, epoxy coated steel, for connection to house exhaust or remote blower
  • Seq:
    17