Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Hard duct connector, epoxy coated steel, for connecting the glove box to ductwork attached to a remote blower (Requires ductwork and remote blower)
Katalog # LBCP5440800
Leverandør:   LABCONCO
Accessories for glove boxes, Protector®
Katalog # LBCP5440800
Leverandør:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Hard duct connector, epoxy coated steel, for connecting the glove box to ductwork attached to a remote blower (Requires ductwork and remote blower)
Price
4.730,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Hard duct connector, epoxy coated steel, for connecting the glove box to ductwork attached to a remote blower (Requires ductwork and remote blower)
  • Seq:
    16