Accessories for glove boxes, Protector®
Hard duct connector, epoxy coated steel, for connecting the glove box to ductwork attached to a remote blower (Requires ductwork and remote blower)
Katalog # LBCP5440800
Leverandør: LABCONCO
Gå til:
Product Details & Documents
Documents
LBCP5242200_TDS_EN
Datablad
Specifikationer
- Beskrivelse:Hard duct connector, epoxy coated steel, for connecting the glove box to ductwork attached to a remote blower (Requires ductwork and remote blower)
- Seq:16