Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Moisture monitor (dew point measurements), 0 - 6000 ppm; incl. meter, probe, attachment plug, and calibration stand; for placement inside the glove box’s main chamber; 140×203×46 mm, 230 V, 50/60 Hz
Catalog # LBCP5442201
Supplier:   LABCONCO
Accessories for glove boxes, Protector®
Catalog # LBCP5442201
Supplier:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Moisture monitor (dew point measurements), 0 - 6000 ppm; incl. meter, probe, attachment plug, and calibration stand; for placement inside the glove box’s main chamber; 140×203×46 mm, 230 V, 50/60 Hz
Price
53.010,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Documents
Specifications
  • Beskrivelse:
    Moisture monitor (dew point measurements), 0 - 6000 ppm; incl. meter, probe, attachment plug, and calibration stand; for placement inside the glove box’s main chamber; 140×203×46 mm, 230 V, 50/60 Hz
  • Seq:
    15