Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Dextran, M.W. ~ 500,000

Glass bottle for solids, 500 g
Katalog # J63702.36
Leverandør:   Thermo Fisher Scientific
Dextran, M.W. ~ 500,000
Katalog # J63702.36
CAS nummer:  9004-54-0
Dextran, M.W. ~ 500,000
Dextran, M.W. ~ 500,000

Glass bottle for solids, 500 g

CAS nummer:  9004-54-0
Price
10.530,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

CAS No.: 9004-54-0 Molecular Formula: (C6H10O5)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,2948 MDL No.: MFCD00130935

  • CAS nummer:9004-54-0
  • Formel:(C₆H₁₀O₅)n
  • MDL nummer:MFCD00130935
  • Merck Index:14,2948
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-677-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer