Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dextran, M.W. ~ 500,000

Dextran, M.W. ~ 500,000

Glass bottle for solids, 50 g
Katalog # J63702.18
Leverandør:   Thermo Fisher Scientific
Dextran, M.W. ~ 500,000
Katalog # J63702.18
CAS nummer:  9004-54-0
Dextran, M.W. ~ 500,000
Dextran, M.W. ~ 500,000

Glass bottle for solids, 50 g

CAS nummer:  9004-54-0
Price
1.840,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

CAS No.: 9004-54-0 Molecular Formula: (C6H10O5)n Physical Form: Powder Appearance: White to off-white Merck Reference: 14,2948 MDL No.: MFCD00130935

  • CAS nummer:9004-54-0
  • Formel:(C₆H₁₀O₅)n
  • MDL nummer:MFCD00130935
  • Merck Index:14,2948
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-677-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.