Forskolin ≥98%
Glass bottle for solids, 10 mg
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 98+% CAS No.: 66575-29-9 Molecular Formula: C22H34O7 Formula Weight: 410.51 Storage Temperature: -30°C to -10°C Physical Form: Solid Solubility: Soluble in DMSO or ethanol. Beilstein Reference: 1692716 Merck Reference: 14,2476 MDL No.: MFCD00082317
An adenylcyclase activator
Synonymer: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
- CAS nummer:66575-29-9
- Formel:C₂₂H₃₄O₇
- MDL nummer:MFCD00082317
- Molecular Weight:410.51 g/mol
- Merck Index:14,2476
- EINECS:266-410-9
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:10 mg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



