Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(S)-(-)-Æblesyre ≥99%

(S)-(-)-Æblesyre ≥99%

Glass bottle for solids, 2,5 kg
Catalog # J63221.A4
Irriterende
(S)-(-)-Æblesyre ≥99%
Catalog # J63221.A4
CAS Number:  97-67-6
(S)-(-)-Æblesyre ≥99%
(S)-(-)-Æblesyre ≥99%

Glass bottle for solids, 2,5 kg

CAS Number:  97-67-6
Price
2.860,00 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents

Purity: 99% CAS No.: 97-67-6 Molecular Formula: C4H6O5 Formula Weight: 134.09 Flash Point: 220°(428°F) Physical Form: Crystalline powder Density: 1.6 Appearance: White Melting Point: 100-106° Beilstein Reference: 1723541 Merck Reference: 14,5707 Optical Rotation: -26° (c=5.5 in pyridine) MDL No.: MFCD00064213

Synonymer:  L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
  • CAS nummer:97-67-6
  • Formel:C₄H₆O₅
  • MDL nummer:MFCD00064213
  • Molecular Weight:134.09 g/mol
  • Merck Index:14,5707
  • Kogende Pt:322 °C (1013 hPa)
  • Smeltende Pt:100…106 °C
  • Tæthed:1,6
  • Flash Pt:220 °C (428 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-601-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2,5 kg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.