Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Ampicillin, natriumsalt

Glass bottle for solids, 5 g
Katalog # J63807.06
Sundhed
Leverandør:   Thermo Fisher Scientific
Ampicillin, natriumsalt
Katalog # J63807.06
CAS nummer:  69-52-3
Ampicillin, natriumsalt
Ampicillin, natriumsalt

Glass bottle for solids, 5 g

CAS nummer:  69-52-3
Price
1.010,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

CAS No.: 69-52-3 Molecular Formula: C16H18N3NaO4S Formula Weight: 371.39 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 215° Solubility: Freely soluble in water. Sparingly soluble in acetone Beilstein Reference: 4119211 Merck Reference: 14,586 MDL No.: MFCD00064313

Synonymer:  D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
  • CAS nummer:69-52-3
  • Formel:C₁₆H₁₈N₃NaO₄S
  • MDL nummer:MFCD00064313
  • Molecular Weight:371.39 g/mol
  • Merck Index:14,586
  • Smeltende Pt:215 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:200-708-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer