Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Polymyxin Bsulfat

Polymyxin Bsulfat

Glass bottle for solids, 1 g
Katalog # J63074.03
Leverandør:   Thermo Fisher Scientific
Polymyxin Bsulfat
Katalog # J63074.03
CAS nummer:  1405-20-5
Polymyxin Bsulfat
Polymyxin Bsulfat

Glass bottle for solids, 1 g

CAS nummer:  1405-20-5
Price
1.640,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

Antibiotic for Gram-negative bacteria.

  • White to pale yellow powder

Soluble in water at 50mg/ml

  • CAS nummer:1405-20-5
  • Formel:C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
  • MDL nummer:MFCD00131991
  • Molecular Weight:1385.61 g/mol
  • Merck Index:14,7573
  • Opbevaringstemperatur:Køleskab
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer