Transfer or Electro Blotting buffer (10X), pH 8,5
Transfer Electroblot Buffer 10X, 8,5, 1 L
Leverandør: Thermo Fisher Scientific
Transfer or Electro Blotting buffer (10X), pH 8,5
Katalog # J63037.K2
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
Physical Form: Liquid
Documents
Specifikationer
Specifikationer
- Pakn:1 L
- Beskrivelse:Transfer Electroblot Buffer 10X
- pH-værdi:8,5
- Cat. No.:J63037.K2
- Physical form:Liquid
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



