Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Luminol and oxidising solutions

Luminol and oxidising solutions

Luminol and oxidising solutions (1 each), 250 mL
Katalog # J61462.AK
Leverandør:   Thermo Fisher Scientific
Luminol and oxidising solutions
Katalog # J61462.AK
CAS nummer:  
Luminol and oxidising solutions

Luminol and oxidising solutions (1 each), 250 mL

CAS nummer:  
Price
1.950,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents

For chemiluminecent HRP-based detection of western blotting systems. It is soluble in most polar organic solvents but is insoluble in water.

Mix equal amounts of luminol and oxidising solution, then incubate the membrane for one minute. Expose membrane to x-ray film for at least one minute. Adjust exposure as necessary.

Applications: For chemiluminecent detection of HRP-based Western blotting systems.

Advarsel: Light Sensitive. Keep Cold.

Documents

Specifikationer

Specifikationer

  • Pakn:
    250 mL
  • Beskrivelse:
    Luminol and oxidising solutions (1 each)

Specifikationer