Orders LinkContactUsLinkComponent
TBE running buffer (10X)
TBE running buffer (10X)
TBE running buffer (10X), 1 L
Catalog # J62788.K2
TBE running buffer (10X)
Catalog # J62788.K2
CAS Number:  
TBE running buffer (10X)
TBE running buffer (10X)
TBE running buffer (10X), 1 L
CAS Number:  
Price
812,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Physical Form: Liquid
Appearance: Colourless
Specifications
Specifications
  • Pakn:
    1 L
  • Beskrivelse:
    TBE running buffer (10X)
  • Cat. No.:
    J62788.K2
  • Udseende:
    Colourless
  • Physical form:
    Liquid
Specifications