Orders LinkContactUsLinkComponent
MES-SDS running buffer (20X)
MES-SDS running buffer (20X), 1 L
Katalog # J62138.K2
Leverandør:   Thermo Fisher Scientific
MES-SDS running buffer (20X)
Katalog # J62138.K2
CAS nummer:  
MES-SDS running buffer (20X)
MES-SDS running buffer (20X)
MES-SDS running buffer (20X), 1 L
CAS nummer:  
Price
2.640,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Specifikationer
Specifikationer
  • Pakn:
    1 L
  • Beskrivelse:
    MES-SDS running buffer (20X)
  • Cat. No.:
    J62138.K2
  • Physical form:
    Liquid
Specifikationer