Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Sodium pyrophosphate, 200 mM buffer solution

Sodium pyrophosphate, 200mM buffer solution, 100 mL
Katalog # J62052.AE
Leverandør:   Thermo Fisher Scientific
Sodium pyrophosphate, 200 mM buffer solution
Katalog # J62052.AE
CAS nummer:  
Sodium pyrophosphate, 200 mM buffer solution
Sodium pyrophosphate, 200 mM buffer solution

Sodium pyrophosphate, 200mM buffer solution, 100 mL

CAS nummer:  
Price
906,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

CAS No.: 7758-16-9
Physical Form: Liquid
MDL No.: MFCD00014246

Specifikationer

Specifikationer

  • Pakn:
    100 mL
  • Beskrivelse:
    Sodium pyrophosphate, 200mM buffer solution
  • Cat. No.:
    J62052.AE
  • CAS-nr:
    7758-16-9
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00014246

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.