Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Neutral rød ACS

Glass bottle for solids, 250 g
Katalog # J62643.30
Leverandør:   Thermo Fisher Scientific
Neutral rød ACS
Katalog # J62643.30
CAS nummer:  553-24-2
Neutral rød ACS
Neutral rød ACS

Glass bottle for solids, 250 g

CAS nummer:  553-24-2
Price
1.540,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents

CAS No.: 553-24-2 Molecular Formula: C15H17ClN4 Formula Weight: 288.78 Physical Form: Powder Appearance: Dark green to black Melting Point: 290° dec. Beilstein Reference: 3918740 Merck Reference: 14,6488 MDL No.: MFCD00012651

Nuclear counterstain useful in pH range 6.8 - 8.0

Synonymer:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS nummer:553-24-2
  • Formel:C₁₅H₁₇ClN₄
  • MDL nummer:MFCD00012651
  • Molecular Weight:288.78 g/mol
  • Merck Index:14,6488
  • Smeltende Pt:290 °C (dec.)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:209-035-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifikationer