Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

U0126 ≥99%

100 mg
Katalog # J61246.MC
Irriterende
Leverandør:   Thermo Fisher Scientific
U0126 ≥99%
Katalog # J61246.MC
CAS nummer:  109511-58-2
U0126 ≥99%
U0126 ≥99%

100 mg

CAS nummer:  109511-58-2
Price
3.900,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2.

  • Soluble in DMSO. Poorly soluble in ethanol and water
  • CAS nummer:109511-58-2
  • Formel:C₁₈H₁₆N₆S₂
  • MDL nummer:MFCD01861186
  • Molecular Weight:380.49 g/mol
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    100 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.