Orders LinkContactUsLinkComponent
U0126 ≥99%
25 mg
  J61246.MB
U0126 ≥99%
  J61246.MB
 :  109511-58-2
U0126 ≥99%
U0126 ≥99%
25 mg
 :  109511-58-2
2.030,00 kr
1 * 25 mg
Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2.
  • Soluble in DMSO. Poorly soluble in ethanol and water
  •  :109511-58-2
  •  :C₁₈H₁₆N₆S₂
  •  :MFCD01861186
  •  :380.49 g/mol
  •  :Fryser
  • Pakn:
    25 mg