U0126 ≥99%
25 mg
Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2.
- Soluble in DMSO. Poorly soluble in ethanol and water
- :109511-58-2
- :C₁₈H₁₆N₆S₂
- :MFCD01861186
- :380.49 g/mol
- :Fryser
- Pakn:25 mg