Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Nystatin

Glass bottle for solids, 5 g
Katalog # J62486.06
Irriterende
Leverandør:   Thermo Fisher Scientific
Nystatin
Katalog # J62486.06
CAS nummer:  1400-61-9
Nystatin
Nystatin

Glass bottle for solids, 5 g

CAS nummer:  1400-61-9
Price
985,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

CAS No.: 1400-61-9 Molecular Formula: C47H75NO17 Formula Weight: 926.09 Storage Temperature: -30°C to -10°C Physical Form: Fine powder Appearance: Off-white to yellowish Melting Point: 160° Solubility: Soluble in DMSO Merck Reference: 14,6736 MDL No.: MFCD00036240

An antimicrobial. Increases the permeability of the cell membrane of sensitive fungi by binding to sterols

Synonymer:  Fungicidin, Mycostatin
  • CAS nummer:1400-61-9
  • Formel:C₄₇H₇₅NO₁₇
  • MDL nummer:MFCD25541651
  • Molecular Weight:926.09 g/mol
  • Merck Index:14,6736
  • Smeltende Pt:160 °C
  • Opbevaringstemperatur:Fryser
  • EINECS:215-749-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer