Buffers, MOPS, 1,0 M solutions
MOPS buffer solution pH 7 (1 mol/l), 7,0, Glass bottle, 500 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL
- Beskrivelse:MOPS buffer solution pH 7 (1 mol/l)
- pH-værdi:7,0
- Cat. No.:J61821.AP
- CAS-nr:1132-61-2
- Udseende:Colourless
- Odour/scent:Odourless
- Physical form:Liquid
- MDL No.:MFCD00006183



