Tyramin ≥98%
Glass bottle for solids, 25 g
Purity: 98+% CAS No.: 51-67-2 Molecular Formula: C8H11NO Formula Weight: 137.18 Physical Form: Powder Appearance: Off-white to yellow Melting Point: 160-162° Boiling Point: 175-181°/8mm Hg Beilstein Reference: 1099914 Merck Reference: 14,9835 MDL No.: MFCD00008193
Reagent for fluorescence enzyme immunoassay
: 4-Hydroxyphenethylamine, 4-(2-Aminoethyl)phenol
- :51-67-2
- :C₈H₁₁NO
- :MFCD00008193
- :137.18 g/mol
- :14,9835
- :175…181 °C (8 mmHg)
- :160…162 °C
- :Stue temperatur
- :200-115-8
- Pakning:Glass bottle for solids
- Pakn:25 g