Orders LinkContactUsLinkComponent
Tyramin ≥98%
Glass bottle for solids, 25 g
  J60990.14
Tyramin ≥98%
  J60990.14
 :  51-67-2
Tyramin ≥98%
Tyramin ≥98%
Glass bottle for solids, 25 g
 :  51-67-2
1.730,00 kr
1 * 25 g
Purity: 98+% CAS No.: 51-67-2 Molecular Formula: C8H11NO Formula Weight: 137.18 Physical Form: Powder Appearance: Off-white to yellow Melting Point: 160-162° Boiling Point: 175-181°/8mm Hg Beilstein Reference: 1099914 Merck Reference: 14,9835 MDL No.: MFCD00008193
Reagent for fluorescence enzyme immunoassay
 :  4-Hydroxyphenethylamine, 4-(2-Aminoethyl)phenol
  •  :51-67-2
  •  :C₈H₁₁NO
  •  :MFCD00008193
  •  :137.18 g/mol
  •  :14,9835
  •  :175…181 °C (8 mmHg)
  •  :160…162 °C
  •  :Stue temperatur
  •  :200-115-8
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g