Orders LinkContactUsLinkComponent
Kinase buffer II (5X)
Kinase buffer II (5X), 100 mL
Catalog # J60269.AE
Kinase buffer II (5X)
Catalog # J60269.AE
CAS Number:  
Kinase buffer II (5X)
Kinase buffer II (5X)
Kinase buffer II (5X), 100 mL
CAS Number:  
Price
670,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents
Specifications
Specifications
  • Pakn:
    100 mL
  • Beskrivelse:
    Kinase buffer II (5X)
  • Cat. No.:
    J60269.AE
  • Physical form:
    Liquid
Specifications