Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tyramin ≥98%

Tyramin ≥98%

Glass bottle for solids, 5 g
Katalog # J60990.06
Irriterende
Leverandør:   Thermo Fisher Scientific
Tyramin ≥98%
Katalog # J60990.06
CAS nummer:  51-67-2
Tyramin ≥98%
Tyramin ≥98%

Glass bottle for solids, 5 g

CAS nummer:  51-67-2
Price
kr510.00
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Purity: 98+% CAS No.: 51-67-2 Molecular Formula: C8H11NO Formula Weight: 137.18 Physical Form: Powder Appearance: Off-white to yellow Melting Point: 160-162° Boiling Point: 175-181°/8mm Hg Beilstein Reference: 1099914 Merck Reference: 14,9835 MDL No.: MFCD00008193

Reagent for fluorescence enzyme immunoassay

Synonymer:  4-Hydroxyphenethylamine, 4-(2-Aminoethyl)phenol
  • CAS nummer:51-67-2
  • Formel:C₈H₁₁NO
  • MDL nummer:MFCD00008193
  • Molecular Weight:137.18 g/mol
  • Merck Index:14,9835
  • Kogende Pt:175…181 °C (8 mmHg)
  • Smeltende Pt:160…162 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-115-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.