5,8,11,14-Eicosatetraynoic acid
Glass bottle for solids, 100 mg
Supplier: Thermo Fisher Scientific
5,8,11,14-Eicosatetraynoic acid
Catalog # J61624.MC
Supplier: Thermo Fisher Scientific
CAS Number: 1191-85-1
Jump to:
Product Details & Documents
CAS No.: 1191-85-1 Molecular Formula: C20H24O2 Formula Weight: 296.40 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Off-white Melting Point: 81-82° Beilstein Reference: 1798411 MDL No.: MFCD00036967
A cyclooxygenase, lipoxygenase and cytochrome P450 inhibitor
- CAS Number:1191-85-1
- Formula:C₂₀H₂₄O₂
- MDL Number:MFCD00036967
- Molecular Weight:296.41 g/mol
- Melting Pt:81…82 °C
Documents
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:100 mg



