Buffers, MOPS, 1,0 M solutions
MOPS buffer solution, 1,0 mol/L, 9,0, Glass bottle, 250 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
CAS No.: 1132-61-2
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00006183
Documents
Specifikationer
Specifikationer
- Koncentration:1,0 mol/L
- Pakning:Glass bottle
- Pakn:250 mL
- Beskrivelse:MOPS buffer solution
- pH-værdi:9,0
- Cat. No.:J60097.AK
- CAS-nr:1132-61-2
- Udseende:Colourless
- Odour/scent:Odourless
- Physical form:Liquid
- MDL No.:MFCD00006183
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



