Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Buffers, MOPS, 0,5 M solutions

MOPS buffer solution, 0,5 mol/L, 7,5, Glass bottle, 22, 100 mL
Katalog # J62839.AE
Leverandør:   Thermo Fisher Scientific
Buffers, MOPS, 0,5 M solutions
Katalog # J62839.AE
CAS nummer:  
Buffers, MOPS, 0,5 M solutions
Buffers, MOPS, 0,5 M solutions

MOPS buffer solution, 0,5 mol/L, 7,5, Glass bottle, 22, 100 mL

CAS nummer:  
Price
888,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Specifikationer

Specifikationer

  • Koncentration:
    0,5 mol/L
  • Pakning:
    Glass bottle
  • Pakn:
    100 mL
  • Beskrivelse:
    MOPS buffer solution
  • Pk Info:
    22
  • pH-værdi:
    7,5
  • Cat. No.:
    J62839.AE
  • CAS-nr:
    1132-61-2
  • Odour/scent:
    Odourless
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00006183
  • Opbevaringstemperatur:
    2°C to 8°C

Specifikationer