Orders LinkContactUsLinkComponent
Brefeldin A ≥99%
Glass bottle for solids, 50 mg
  J62340.MF
Brefeldin A ≥99%
  J62340.MF
 :  20350-15-6
Brefeldin A ≥99%
Brefeldin A ≥99%
Glass bottle for solids, 50 mg
 :  20350-15-6
7.600,00 kr
1 * 50 mg
 :  (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
  •  :20350-15-6
  •  :C₁₆H₂₄O₄
  •  :MFCD00083258
  •  :280.4 g/mol
  •  :14,1369
  •  :201…205 °C
  •  :Fryser
  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 mg