Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Brefeldin A ≥99%

Brefeldin A ≥99%

Glass bottle for solids, 25 mg
Katalog # J62340.MB
Irriterende
Leverandør:   Thermo Fisher Scientific
Brefeldin A ≥99%
Katalog # J62340.MB
CAS nummer:  20350-15-6
Brefeldin A ≥99%
Brefeldin A ≥99%

Glass bottle for solids, 25 mg

CAS nummer:  20350-15-6
Price
4.530,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents
Synonymer:  (1R,2E,6S,10E,11aS,13S,14aR)-1,13-dihydroxy-6-methyl-6,7,8,9,12,13,14,14a-octahydro-1H-cyclopenta[f][1]oxacyclotridecin-4(11aH)-one, Brefeldin A, natural
  • CAS nummer:20350-15-6
  • Formel:C₁₆H₂₄O₄
  • MDL nummer:MFCD00083258
  • Molecular Weight:280.4 g/mol
  • Merck Index:14,1369
  • Smeltende Pt:201…205 °C
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 mg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.