Orders LinkContactUsLinkComponent
MNE buffer, pH 6,5
MNE buffer, pH 6.5, 500 mL
Katalog # J60690.AP
Leverandør:   Thermo Fisher Scientific
MNE buffer, pH 6,5
Katalog # J60690.AP
CAS nummer:  
MNE buffer, pH 6,5
MNE buffer, pH 6,5
MNE buffer, pH 6.5, 500 mL
CAS nummer:  
Price
745,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Specifikationer
Specifikationer
  • Pakn:
    500 mL
  • Beskrivelse:
    MNE buffer, pH 6.5
  • Cat. No.:
    J60690.AP
  • Physical form:
    Liquid