Orders LinkContactUsLinkComponent
Acrylamide/Bis-Acrylamide
Acrylamide/Bis-Acrylamide
Acrylamide/Bisacrylamide, (37,5:1), 30% solution, colourless
Catalog # J61505.K2
Acrylamide/Bis-Acrylamide
Catalog # J61505.K2
Acrylamide/Bis-Acrylamide
Acrylamide/Bis-Acrylamide
Acrylamide/Bisacrylamide, (37,5:1), 30% solution, colourless
Price
2.950,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Premix of acrylamide with the common cross-linking reagent of final ratio 29:1.
  • Soluble in water
  • DNA sequencing gels and protein separation
Specifications
  • Beskrivelse:
    Acrylamide/Bisacrylamide, (37,5:1), 30% solution, colourless
  • Seq:
    02