MOPSO, 0,2 M buffer soln., pH 7,0
MOPSO buffer solution, 0,2 M, 7,0, 250 mL
Leverandør: Thermo Fisher Scientific
MOPSO, 0,2 M buffer soln., pH 7,0
Katalog # J61469.AK
Leverandør: Thermo Fisher Scientific
CAS nummer:
Gå til:
Product Details & Documents
CAS No.: 79803-73-9
Physical Form: Liquid
MDL No.: MFCD00070007
Specifikationer
Specifikationer
- Koncentration:0,2 M
- Pakn:250 mL
- Beskrivelse:MOPSO buffer solution
- pH-værdi:7,0
- Cat. No.:J61469.AK
- CAS-nr:79803-73-9
- Physical form:Liquid
- MDL No.:MFCD00070007



