Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Inoculating loops, Microloops® Nichrome 5

Inoculating loops, NiCr, 3,3 µl, 2,9 mm
  MEDWMW192
Inoculating loops, Microloops® Nichrome 5
  MEDWMW192
Inoculating loops, Microloops® Nichrome 5

Inoculating loops, NiCr, 3,3 µl, 2,9 mm

 
1.060,00 kr
   
1 * 25 Items
 

Microloops® are a range of inoculating loops that have been designed and moulded with precision, to ensure you can rely on them for accurate volumetric technique every time.

  • Twisted wire - reduces vibrations
  • Stable during repeated heat/cool cycles
  • Fully closed loop for secure transfer of drop

Twisted Nichrome 5 wire loops offer all the benefits of platinum, without the costs. These are available in different volumetric sizes. Nichrome 5 is the finest for repeated heating. It is made up of 80% Nickel and 20% Chromium. It
gives outstanding performance at temperatures up to 1200 °C.

 
  • Ø:
    2,9 mm
  • Kapacitet:
    3,3 µl
  • Beskrivelse:
    Inoculating loops, NiCr
  • Seq:
    04