Orders LinkContactUsLinkComponent
Accessories for Loopster basic and digital
Tube attachment for 20×15 ml tubes with Ø16 mm
  444-0852
 :   IKA
Accessories for Loopster basic and digital
  444-0852
 :  IKA
Accessories for Loopster basic and digital
Accessories for Loopster basic and digital
Tube attachment for 20×15 ml tubes with Ø16 mm
1.470,00 kr
1 * 1 Items
  • Beskrivelse:
    Tube attachment for 20×15 ml tubes with Ø16 mm
  • Seq:
    05
  • Cat. No.:
    444-0852
Accessories
  •  
    Beskrivelse
  • 444-0952
    Spare clips with Ø25 mm for use with Universal disc
    1 * 12 Items - 384,00 kr
  •  
    Beskrivelse
  • 444-0857
    Spare clips with Ø12 mm for use with Universal disc
    1 * 12 Items - 384,00 kr
  •  
    Beskrivelse
  • 444-0856
    Spare clips with Ø10 mm for use with Universal disc
    1 * 12 Items - 384,00 kr
  •  
    Beskrivelse
  • 444-0953
    Spare clips with Ø28 mm for use with Universal disc
    1 * 12 Items - 384,00 kr
  •  
    Beskrivelse
  • 444-0951
    Spare clips with Ø16 mm for use with Universal disc
    1 * 12 Items - 384,00 kr