Orders LinkContactUsLinkComponent
Accessories for Loopster basic and digital
Accessories for Loopster basic and digital
Spare clips with Ø12 mm for use with Universal disc
Katalog # 444-0857
Leverandør:   IKA
Accessories for Loopster basic and digital
Katalog # 444-0857
Leverandør:  IKA
Accessories for Loopster basic and digital
Accessories for Loopster basic and digital
Spare clips with Ø12 mm for use with Universal disc
Price
384,00 kr
Log ind for tilgængelighed
1 * 12 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Spare clips with Ø12 mm for use with Universal disc
  • Seq:
    10
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Spare clips with Ø25 mm for use with Universal disc
    Spare clips with Ø25 mm for use with Universal disc
    444-0952
    Log ind for tilgængelighed
    1 * 12 Items - 384,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Spare clips with Ø12 mm for use with Universal disc
    Spare clips with Ø12 mm for use with Universal disc
    444-0857
    Log ind for tilgængelighed
    1 * 12 Items - 384,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Spare clips with Ø10 mm for use with Universal disc
    Spare clips with Ø10 mm for use with Universal disc
    444-0856
    Log ind for tilgængelighed
    1 * 12 Items - 384,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Spare clips with Ø28 mm for use with Universal disc
    Spare clips with Ø28 mm for use with Universal disc
    444-0953
    Log ind for tilgængelighed
    1 * 12 Items - 384,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Spare clips with Ø16 mm for use with Universal disc
    Spare clips with Ø16 mm for use with Universal disc
    444-0951
    Log ind for tilgængelighed
    1 * 12 Items - 384,00 kr