Stirrer blades
Anchor stirrer blades, PTFE, with stiffened metal core, for SVL reaction vessels, 54 mm
Katalog # BIBB588-98
Leverandør: SciLabware
Gå til:
Product Details & Documents
A range of PTFE coated stirrer heads designed to screw into the threads on SVL® glass stirrer shafts.
- Head simply screws into the end of the shaft
- Anchor and helix types have a PTFE coated metal core
Advarsel: Stirrers must be rotated in a clockwise direction to prevent unscrewing of the thread.
Specifikationer
- Model:Anchor stirrer blades, PTFE, with stiffened metal core, for SVL reaction vessels
- Bredde:54 mm
- Seq:012