Orders LinkContactUsLinkComponent
Powder funnels, PP/PE
Powder funnels, PP/PE
150 mm, 28 mm
Katalog # BURK9606-0150
Leverandør:   BURKLE
Powder funnels, PP/PE
Katalog # BURK9606-0150
Leverandør:  BURKLE
Powder funnels, PP/PE
Powder funnels, PP/PE
150 mm, 28 mm
Price
71,10 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These powder funnels are designed for safe filling of containers.
  • PP/PE
9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
Specifikationer
  • Tragt udv. Ø:
    150 mm
  • Seq:
    03
  • Stilk udv. Ø:
    28 mm