Orders LinkContactUsLinkComponent
Tilbehør
Round vials with lid , height 48 mm, Ø 24 mm
Catalog # LOVI197629
Supplier:   Lovibond
Tilbehør
Catalog # LOVI197629
Supplier:  Lovibond
Tilbehør
Tilbehør
Round vials with lid , height 48 mm, Ø 24 mm
Price
497,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
Specifications
  • Beskrivelse:
    Round vials with lid , height 48 mm, Ø 24 mm