Orders LinkContactUsLinkComponent
CF® Dye Azide
CF® Dye Azide
CF®405M Azide, 408/452 nm, 0,5 mg
Katalog # 92092.
Leverandør:   Biotium
CF® Dye Azide
Katalog # 92092.
Leverandør:  Biotium
CAS nummer:  
CF® Dye Azide
CF® Dye Azide
CF®405M Azide, 408/452 nm, 0,5 mg
CAS nummer:  
Price
3.650,00 kr
Log ind for tilgængelighed
1 * 0,5 mg
Product Details & Documents
CF® Dyes with an azide group. Can be used to label or detect alkyne, BCN or cyclooctyne-containing molecules by fluorescence spectroscopy, microscopy or flow cytometry following copper-catalyzed azide-alkyne cycloaddition.
  • Fluorescent detection or labeling of alkyne, BCN or cyclooctyne groups on target molecules
  • Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labeling
CF® Dye azide can react with terminal alkynes to form 1,2,3-triazole via copper-catalyzed cycloaddition. They can also undergo Staudinger ligation with a phosphine containing compounds. The azide derivatives also react with cyclooctynes and BCN groups via copper-free bioorthogonal reaction.
Superior CF® Dyes, CF® Dyes are Biotium’s line of next-generation fluorescent dyes that have improved brightness, photostability and water solubility compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Specifikationer
Specifikationer
  • Pakn:
    0,5 mg
  • Beskrivelse:
    CF®405M Azide
  • Ex/Em:
    408/452 nm