Product Details & Documents
CF® Dyes with an azide group. Can be used to label or detect alkyne, BCN or cyclooctyne-containing molecules by fluorescence spectroscopy, microscopy or flow cytometry following copper-catalyzed azide-alkyne cycloaddition.
- Fluorescent detection or labeling of alkyne, BCN or cyclooctyne groups on target molecules
- Bright, photostable and water-soluble CF® Dyes are excellent options for fluorescent labeling
CF® Dye azide can react with terminal alkynes to form 1,2,3-triazole via copper-catalyzed cycloaddition. They can also undergo Staudinger ligation with a phosphine containing compounds. The azide derivatives also react with cyclooctynes and BCN groups via copper-free bioorthogonal reaction.
Superior CF® Dyes, CF® Dyes are Biotium’s line of next-generation fluorescent dyes that have improved brightness, photostability and water solubility compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Specifikationer
Specifikationer
- Pakn:0,5 mg
- Beskrivelse:CF®647 Azide
- Ex/Em:650/665 nm