Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Filter Holder Accessories

Filter Holder Accessories

Vent/relief valve, 9 bar, Int.Ø 6,3 mm, 293 mm stainless steel sanitary filter holder
PL-1 cartridge housings
for use with PS-1 cartridge housings
Katalog # MILFXX6700024
Leverandør:   Merck
Filter Holder Accessories
Katalog # MILFXX6700024
Leverandør:  Merck
Filter Holder Accessories
Filter Holder Accessories

Vent/relief valve, 9 bar, Int.Ø 6,3 mm, 293 mm stainless steel sanitary filter holder
PL-1 cartridge housings
for use with PS-1 cartridge housings

Price
2.320,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Til:
    293 mm stainless steel sanitary filter holder
    PL-1 cartridge housings
    for use with PS-1 cartridge housings
  • Beskrivelse:
    Vent/relief valve, 9 bar, Int.Ø 6,3 mm
  • Seq:
    27