HPLC columns, Acclaim™ Trinity™ Q1
Beskyttelsespatron, 300 Å, 3 µm, 3,00 mm, 10 mm
Katalog # 555-7530
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
The Acclaim™ Trinity™ Q1 column is a mixed-mode, silica-based application-specific column for high-resolution and high-throughput trace analysis of herbicides diquat and paraquat by LC-MS/MS and LC-UV methods.
- No need for ion-pairing reagent use
- LC/MS compatible
- Direct injection possible with on-line SPE
- Excellent resolution, good peak shape
Particle type: ultra pure, spherical silica
Bonded phase: mixed-mode reversed phase, anion exchange and cation exchange
pH range: 2,5 to 7,5
Specifikationer
- Kolonne form:Beskyttelsespatron
- Ø indv.:3,00 mm
- Partikelstørrelse:3 µm
- Porestørrelse:300 Å
- Længde:10 mm
- Seq:01
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Stainless steel guard cartridge holder, V-2Stainless steel guard cartridge holder, V-2554-4528Log ind for tilgængelighed1 * 1 Items - 3.610,00 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



