HPLC columns, Acclaim™ Trinity™ Q1
Analytisk, 300 Å, 3 µm, 3,00 mm, 50 mm
Catalog # 555-7384
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
The Acclaim™ Trinity™ Q1 column is a mixed-mode, silica-based application-specific column for high-resolution and high-throughput trace analysis of herbicides diquat and paraquat by LC-MS/MS and LC-UV methods.
- No need for ion-pairing reagent use
- LC/MS compatible
- Direct injection possible with on-line SPE
- Excellent resolution, good peak shape
Particle type: ultra pure, spherical silica
Bonded phase: mixed-mode reversed phase, anion exchange and cation exchange
pH range: 2,5 to 7,5
Specifications
- Kolonne form:Analytisk
- Ø indv.:3,00 mm
- Partikelstørrelse:3 µm
- Porestørrelse:300 Å
- Længde:50 mm
- Seq:02
Accessories
Product Information
- Catalog #BeskrivelseAvailabilityPrice
- 554-4528Stainless steel guard cartridge holder, V-2Log in for availability1 * 1 Items - 3.610,00 kr



