Orders LinkContactUsLinkComponent
Bag cutters
Bag cutters
Irradiated bag cutter, sterile, indivually packed
Catalog # 811-0165
Supplier:   MICRONOVA
Bag cutters
Catalog # 811-0165
Supplier:  MICRONOVA
Bag cutters
Bag cutters
Irradiated bag cutter, sterile, indivually packed
Price
7.210,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
Plastic cutters for opening raw material packages.
  • Automatically retractable safety blade guard
  • No blade changes required - eliminates all loose blades
  • Ergonomic handle
  • Available gamma irradiated
Specifications
  • Beskrivelse:
    Irradiated bag cutter, sterile, indivually packed
  • Seq:
    02