Waste caps
Waste cap, includes O-ring, 1/16" and 1/8" nuts, ferrules and colour-sleeve adapter, GL 45, 3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 10M x 1 thread for barbed adapter, PP
JOURJR-S-11023
These waste caps include three or four ports for ¹/₈ and ¹/₁₆" tubing. Two versions of each waste cap are available, with an extra port to accommodate either a 1/2-20 port for ¹/₄" O.Ø tubing, or a barbed hose adapter.
: Barbed adapters must be ordered separately.
- Gevind:GL 45
- Beskrivelse:Waste cap, includes O-ring, 1/16" and 1/8" nuts, ferrules and colour-sleeve adapter
- Materiale:PP
- Seq:05
- Stik:3 ports with 1/4-28 thread for 1/8" or 1/16" tubing, 1 port with 10M x 1 thread for barbed adapter